Assay Details
| Gene Symbol: HFR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331332.1 | NP_001323315.1 | 2 - 3 | 815 | 66 | |
| NM_100115.3 | NP_563650.1 | 3 - 4 | 691 | 66 | ||
| GenBank mRNA | AF288287.1 | - | 2 - 3 | 496 | 66 | |
| AF323182.1 | - | 3 - 4 | 655 | 66 | ||
| AF324245.1 | - | 3 - 4 | 570 | 66 | ||
More Information
Set Membership: |
|

HFR1
Genomic Map