Assay Details
| Gene Symbol: UBP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112954.4 | NP_566666.2 | 12 - 13 | 1641 | 135 | |
| GenBank mRNA | AK226894.1 | - | 12 - 13 | 1491 | 135 | |
| AX765820.1 | - | 12 - 13 | 1434 | 135 | ||
| AY140096.1 | - | 12 - 13 | 1496 | 135 | ||
More Information
Set Membership: |
|

UBP14
Genomic Map