Assay Details
| Gene Symbol: UBP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112954.4 | NP_566666.2 | 14 - 15 | 1828 | 82 | |
| GenBank mRNA | AK226894.1 | - | 14 - 15 | 1678 | 82 | |
| AX765820.1 | - | 14 - 15 | 1621 | 82 | ||
| AY140096.1 | - | 14 - 15 | 1683 | 82 | ||
More Information
Set Membership: |
|

UBP14
Genomic Map