Assay Details
| Gene Symbol: UBP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112954.4 | NP_566666.2 | 16 - 17 | 2011 | 78 | |
| GenBank mRNA | AK226894.1 | - | 16 - 17 | 1861 | 78 | |
| AX765820.1 | - | 16 - 17 | 1804 | 78 | ||
| AY140096.1 | - | 16 - 17 | 1866 | 78 | ||
More Information
Set Membership: |
|

UBP14
Genomic Map