Assay Details
| Gene Symbol: AT3G20800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112971.4 | NP_188716.1 | 2 - 3 | 366 | 70 | |
| GenBank mRNA | AY091082.1 | - | 2 - 3 | 297 | 70 | |
| AY122953.1 | - | 2 - 3 | 250 | 70 | ||
More Information
Set Membership: |
|

AT3G20800
Genomic Map