Assay Details
| Gene Symbol: PIP1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130159.4 | NP_182120.1 | 3 - 4 | 1141 | 80 | |
| GenBank mRNA | AY048294.1 | - | 3 - 4 | 832 | 80 | |
| AY063931.1 | - | 3 - 4 | 832 | 80 | ||
| AY091254.1 | - | 3 - 4 | 772 | 80 | ||
| HM217349.1 | - | 3 - 4 | 809 | 80 | ||
| X68293.1 | - | 826 | 80 | |||
More Information
Set Membership: |
|

PIP1B
Genomic Map