Assay Details
| Gene Symbol: ACLA-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100814.4 | NP_172414.1 | 10 - 11 | 1220 | 89 | |
| GenBank mRNA | AY127019.1 | - | 10 - 11 | 1112 | 89 | |
| BT003017.1 | - | 10 - 11 | 1081 | 89 | ||
More Information
Set Membership: |
|

ACLA-3
Genomic Map