Assay Details
| Gene Symbol: GPAT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125455.4 | NP_568925.1 | 4 - 5 | 522 | 90 | |
| GenBank mRNA | AK118869.1 | - | 4 - 5 | 448 | 90 | |
| BT006033.1 | - | 4 - 5 | 398 | 90 | ||
| FJ479752.1 | - | 4 - 5 | 398 | 90 | ||
More Information
Set Membership: |
|

GPAT9
Genomic Map