Assay Details
| Gene Symbol: COBL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125488.5 | NP_200903.1 | 1 - 2 | 217 | 88 | |
| GenBank mRNA | BT010831.1 | - | 1 - 2 | 150 | 88 | |
| BT011296.1 | - | 1 - 2 | 71 | 88 | ||
More Information
Set Membership: |
|

COBL5
Genomic Map