Assay Details
| Gene Symbol: AT1G49030 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103796.4 | NP_175332.1 | 1 - 2 | 430 | 83 | |
| GenBank mRNA | BT010928.1 | - | 1 - 2 | 333 | 83 | |
| BT011646.1 | - | 1 - 2 | 302 | 83 | ||
More Information
Set Membership: |
|

AT1G49030
Genomic Map