Assay Details
| Gene Symbol: ETFALPHA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103974.6 | NP_175507.1 | 3 - 4 | 566 | 107 | |
| GenBank mRNA | AK117515.1 | - | 3 - 4 | 524 | 107 | |
| BT004989.1 | - | 3 - 4 | 493 | 107 | ||
More Information
Set Membership: |
|

ETFALPHA
Genomic Map