Assay Details
| Gene Symbol: CID7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336049.1 | NP_001324897.1 | 5 - 6 | 1683 | 68 | |
| NM_001336050.1 | NP_001324896.1 | 5 - 6 | 1683 | 68 | ||
| NM_001336051.1 | NP_001324893.1 | 4 - 5 | 1761 | 68 | ||
| NM_001336052.1 | NP_001324892.1 | 5 - 6 | 1531 | 68 | ||
| NM_128185.3 | NP_180196.1 | 5 - 6 | 1542 | 68 | ||
| GenBank mRNA | BT030338.1 | - | 3 - 4 | 1284 | 68 | |
More Information
Set Membership: |
|

CID7
Genomic Map