Assay Details
| Gene Symbol: SMU2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128203.2 | NP_180214.1 | 5 - 6 | 561 | 98 | |
| GenBank mRNA | BT011249.1 | - | 5 - 6 | 457 | 98 | |
| BT012549.1 | - | 5 - 6 | 404 | 98 | ||
More Information
Set Membership: |
|

SMU2
Genomic Map