Assay Details
| Gene Symbol: AT3G21770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113072.2 | NP_188814.1 | 2 - 3 | 481 | 85 | |
| GenBank mRNA | AY072326.1 | - | 2 - 3 | 481 | 85 | |
| AY114567.1 | - | 2 - 3 | 412 | 85 | ||
More Information
Set Membership: |
|

AT3G21770
Genomic Map