Assay Details
| Gene Symbol: AT1G12790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101149.4 | NP_563913.1 | 8 - 9 | 911 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G12790| Gene Symbol: AT1G12790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101149.4 | NP_563913.1 | 8 - 9 | 911 | 94 | |
Set Membership: |
|