Assay Details
| Gene Symbol: LAX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106418.4 | NP_177892.1 | 4 - 5 | 757 | 64 | |
| GenBank mRNA | AY054266.1 | - | 4 - 5 | 679 | 64 | |
| BT008300.1 | - | 4 - 5 | 656 | 64 | ||
More Information
Set Membership: |
|

LAX3
Genomic Map