Assay Details
| Gene Symbol: VHA-A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036222.3 | NP_001031299.1 | 1 - 2 | 347 | 63 | |
| NM_106539.6 | NP_178011.1 | 2 - 3 | 306 | 63 | ||
| GenBank mRNA | AK230248.1 | - | 2 - 3 | 575 | 63 | |
| AK317358.1 | - | 1 - 2 | 304 | 63 | ||
| AY059909.1 | - | 2 - 3 | 207 | 63 | ||
| AY081296.1 | - | 2 - 3 | 207 | 63 | ||
| BT002589.1 | - | 1 - 2 | 82 | 63 | ||
| BT008383.1 | - | 1 - 2 | 82 | 63 | ||
| U65638.1 | - | 1 - 2 | 110 | 63 | ||
| Gene Symbol: AT5G21020 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK230328.1 | - | 2154 | 63 | ||
More Information
Set Membership: |
|

VHA-A
Genomic Map