Assay Details
| Gene Symbol: HAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198504.2 | NP_001185433.1 | 15 - 16 | 4788 | 142 | |
| NM_001334866.1 | NP_001322202.1 | 15 - 16 | 4664 | 142 | ||
| NM_106550.4 | NP_565197.3 | 15 - 16 | 4904 | 142 | ||
More Information
Set Membership: |
|

HAC1
Genomic Map