Assay Details
| Gene Symbol: NUDT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334912.1 | NP_001320544.1 | 10 - 11 | 1144 | 78 | |
| NM_106618.5 | NP_565218.1 | 9 - 10 | 1316 | 78 | ||
| GenBank mRNA | AY120774.1 | - | 9 - 10 | 1032 | 78 | |
| BT000142.1 | - | 9 - 10 | 1002 | 78 | ||
More Information
Set Membership: |
|

NUDT3
Genomic Map