Assay Details
| Gene Symbol: NIP5;1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117106.3 | NP_192776.1 | 2 - 3 | 1540 | 103 | |
| GenBank mRNA | AY087560.1 | - | 2 - 3 | 966 | 103 | |
| BT030327.1 | - | 2 - 3 | 654 | 103 | ||
More Information
Set Membership: |
|

NIP5;1
Genomic Map