Assay Details
| Gene Symbol: RBL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198164.2 | NP_001185093.1 | 2 - 3 | 544 | 75 | |
| NM_102339.3 | NP_173900.2 | 2 - 3 | 460 | 75 | ||
More Information
Set Membership: |
|

RBL10
Genomic Map