Assay Details
| Gene Symbol: VTI1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102430.5 | NP_564255.1 | 1 - 2 | 432 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

VTI1B| Gene Symbol: VTI1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102430.5 | NP_564255.1 | 1 - 2 | 432 | 90 | |
Set Membership: |
|