Assay Details
| Gene Symbol: AGO10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203535.1 | NP_001190464.1 | 8 - 9 | 1897 | 91 | |
| NM_001344505.1 | NP_001331600.1 | 7 - 8 | 1820 | 91 | ||
| NM_001344506.1 | NP_001331599.1 | 7 - 8 | 1637 | 91 | ||
| NM_123748.3 | NP_199194.1 | 7 - 8 | 1692 | 91 | ||
More Information
Set Membership: |
|

AGO10
Genomic Map