Assay Details
| Gene Symbol: AGO10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203535.1 | NP_001190464.1 | 10 - 11 | 2120 | 119 | |
| NM_001344505.1 | NP_001331600.1 | 9 - 10 | 2043 | 119 | ||
| NM_001344506.1 | NP_001331599.1 | 9 - 10 | 1860 | 119 | ||
| NM_123748.3 | NP_199194.1 | 9 - 10 | 1915 | 119 | ||
More Information
Set Membership: |
|

AGO10
Genomic Map