Assay Details
| Gene Symbol: RPS19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124103.4 | NP_568681.1 | 4 - 5 | 482 | 84 | |
| GenBank mRNA | AY045963.1 | - | 4 - 5 | 382 | 84 | |
| AY091354.1 | - | 3 - 4 | 285 | 84 | ||
More Information
Set Membership: |
|

RPS19
Genomic Map