Assay Details
| Gene Symbol: AT5G51400 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124519.4 | NP_568759.1 | 3 - 4 | 375 | 83 | |
| GenBank mRNA | AY054680.1 | - | 3 - 4 | 309 | 83 | |
| BT002573.1 | - | 2 - 3 | 227 | 83 | ||
| U78870.1 | - | 3 - 4 | 325 | 83 | ||
More Information
Set Membership: |
|

AT5G51400
Genomic Map