Assay Details
| Gene Symbol: GAUT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202773.2 | NP_001189702.1 | 2 - 3 | 448 | 96 | |
| NM_001336714.1 | NP_001323491.1 | 2 - 3 | 427 | 96 | ||
| NM_129422.4 | NP_565893.1 | 2 - 3 | 528 | 96 | ||
| GenBank mRNA | AY050982.1 | - | 2 - 3 | 372 | 96 | |
| AY091448.1 | - | 2 - 3 | 175 | 96 | ||
| KJ138859.1 | - | 2 - 3 | 175 | 96 | ||
More Information
Set Membership: |
|

GAUT7
Genomic Map