Assay Details
| Gene Symbol: AT3G10980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111935.6 | NP_187709.5 | 1 | 2549 | 85 | |
| GenBank mRNA | AY062465.1 | - | 1 | 2402 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT3G10980| Gene Symbol: AT3G10980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111935.6 | NP_187709.5 | 1 | 2549 | 85 | |
| GenBank mRNA | AY062465.1 | - | 1 | 2402 | 85 | |
Set Membership: |
|