Assay Details
| Gene Symbol: TUA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121983.4 | NP_197479.1 | 1 - 2 | 290 | 171 | |
| Gene Symbol: TUA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY040012.1 | - | 1 - 2 | 225 | 171 | |
| AY079366.1 | - | 1 - 2 | 108 | 171 | ||
More Information
Set Membership: |
|

TUA5
Genomic Map