Assay Details
| Gene Symbol: AT5G38030 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123162.4 | NP_198619.1 | 3 - 4 | 1192 | 149 | |
| GenBank mRNA | AF372972.1 | - | 3 - 4 | 995 | 149 | |
| BT002707.1 | - | 3 - 4 | 928 | 149 | ||
More Information
Set Membership: |
|

AT5G38030
Genomic Map