Assay Details
| Gene Symbol: BZIP17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129659.3 | NP_565946.1 | 1 - 2 | 1967 | 145 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BZIP17| Gene Symbol: BZIP17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129659.3 | NP_565946.1 | 1 - 2 | 1967 | 145 | |
Set Membership: |
|