Assay Details
| Gene Symbol: MEF14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113590.4 | NP_189313.2 | 1 - 2 | 655 | 141 | |
| GenBank mRNA | AK117416.1 | - | 1 - 2 | 559 | 141 | |
More Information
Set Membership: |
|

MEF14
Genomic Map