Assay Details
| Gene Symbol: LSM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101177.1 | NP_001094647.1 | 3 - 4 | 304 | 97 | |
| GenBank mRNA | BC150005.1 | - | 3 - 4 | 304 | 97 | |
More Information
Set Membership: |
|

LSM2
Genomic Map