Assay Details
| Gene Symbol: ENTPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174536.2 | NP_776961.1 | 2 - 3 | 211 | 67 | |
| GenBank mRNA | AF005940.1 | - | 2 - 3 | 211 | 67 | |
| BC109518.1 | - | 2 - 3 | 322 | 67 | ||
More Information
Set Membership: |
|

ENTPD1
Genomic Map