Assay Details
| Gene Symbol: FUT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176851.1 | NP_789821.1 | 3 - 4 | 306 | 62 | |
| GenBank mRNA | AJ132773.2 | - | 3 - 4 | 306 | 62 | |
| AJ132774.1 | - | 1 - 2 | 77 | 62 | ||
| AJ132775.1 | - | 1 - 2 | 52 | 62 | ||
| AJ132776.1 | - | 1 - 2 | 46 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ132773.2, AJ132774.1, AJ132775.1, AJ132776.1, NM_176851.1 transcripts.
Set Membership: |
|

FUT6
Genomic Map