Assay Details
| Gene Symbol: PPARA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034036.1 | NP_001029208.1 | 4 - 5 | 590 | 61 | |
| GenBank mRNA | AF229356.1 | - | 2 - 3 | 363 | 61 | |
| BT020756.1 | - | 4 - 5 | 590 | 61 | ||
More Information
Set Membership: |
|

PPARA
Genomic Map