Assay Details
| Gene Symbol: ASB14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035070.2 | NP_001030242.2 | 9 - 10 | 1618 | 68 | |
| GenBank mRNA | BC148874.1 | - | 9 - 10 | 1619 | 68 | |
| BT021867.1 | - | 4 - 5 | 1047 | 68 | ||
More Information
Set Membership: |
|

ASB14
Genomic Map