Assay Details
| Gene Symbol: CALB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035288.1 | NP_001030365.1 | 4 - 5 | 416 | 57 | |
| GenBank mRNA | BC103038.1 | - | 4 - 5 | 416 | 57 | |
More Information
Set Membership: |
|

CALB2
Genomic Map