Assay Details
| Gene Symbol: VDAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174485.4 | NP_776910.2 | 6 - 7 | 624 | 66 | |
| GenBank mRNA | AF268464.1 | - | 5 - 6 | 546 | 66 | |
| BC102113.1 | - | 6 - 7 | 626 | 66 | ||
More Information
Set Membership: |
|

VDAC1
Genomic Map