Assay Details
| Gene Symbol: CKM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174773.4 | NP_777198.2 | 6 - 7 | 853 | 65 | |
| GenBank mRNA | AF120106.1 | - | 5 - 6 | 778 | 65 | |
| BC102907.1 | - | 6 - 7 | 854 | 65 | ||
| BT020785.1 | - | 6 - 7 | 836 | 65 | ||
| BT021173.1 | - | 6 - 7 | 839 | 65 | ||
More Information
Set Membership: |
|

CKM
Genomic Map