Assay Details
| Gene Symbol: GCNT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205809.1 | NP_991378.1 | 1 - 2 | 123 | 81 | |
| GenBank mRNA | AY283763.1 | - | 1 - 2 | 123 | 81 | |
| AY283764.1 | - | 1 - 2 | 123 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_205809.1 transcripts.
Set Membership: |
|

GCNT3
Genomic Map