Assay Details
| Gene Symbol: PGAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034054.1 | NP_001029226.1 | 1 - 2 | 151 | 113 | |
| GenBank mRNA | AB098929.1 | - | 1 - 2 | 206 | 113 | |
| BC103115.1 | - | 1 - 2 | 171 | 113 | ||
More Information
Set Membership: |
|

PGAM1
Genomic Map