Assay Details
| Gene Symbol: CCL19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034345.1 | NP_001029517.1 | 1 - 2 | 185 | 91 | |
| GenBank mRNA | BC103203.1 | - | 1 - 2 | 185 | 91 | |
More Information
Set Membership: |
|

CCL19
Genomic Map