Assay Details
| Gene Symbol: ISCA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034470.2 | NP_001029642.1 | 2 - 3 | 220 | 87 | |
| GenBank mRNA | BC102863.1 | - | 2 - 3 | 221 | 87 | |
More Information
Set Membership: |
|

ISCA1
Genomic Map