Assay Details
| Gene Symbol: GPRC5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034515.1 | NP_001029687.1 | 3 - 4 | 1111 | 129 | |
| GenBank mRNA | BC105147.1 | - | 3 - 4 | 1111 | 129 | |
More Information
Set Membership: |
|

GPRC5A
Genomic Map