Assay Details
| Gene Symbol: AMY2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035016.1 | NP_001030188.1 | 3 - 4 | 607 | 150 | |
| GenBank mRNA | BC105233.1 | - | 3 - 4 | 607 | 150 | |
More Information
Set Membership: |
|

AMY2B
Genomic Map