Assay Details
| Gene Symbol: SDCBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075483.2 | NP_001068951.1 | 7 - 8 | 834 | 133 | |
| GenBank mRNA | BC103011.1 | - | 7 - 8 | 835 | 133 | |
| BT030535.1 | - | 8 - 9 | 880 | 133 | ||
More Information
Set Membership: |
|

SDCBP
Genomic Map