Assay Details
| Gene Symbol: ZNF33B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075810.1 | NP_001069278.1 | 2 - 3 | 224 | 71 | |
| GenBank mRNA | BC104558.1 | - | 2 - 3 | 224 | 71 | |
More Information
Set Membership: |
|

ZNF33B
Genomic Map