Assay Details
| Gene Symbol: CXCL13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015576.2 | NP_001015576.1 | 1 - 2 | 110 | 99 | |
| GenBank mRNA | AY911364.1 | - | 1 - 2 | 111 | 99 | |
| BC109998.1 | - | 1 - 2 | 110 | 99 | ||
More Information
Set Membership: |
|

CXCL13
Genomic Map