Assay Details
| Gene Symbol: C19H17orf49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038092.1 | NP_001033181.1 | 2 - 3 | 184 | 70 | |
| GenBank mRNA | BC109638.1 | - | 2 - 3 | 184 | 70 | |
More Information
Set Membership: |
|

C19H17orf49
Genomic Map